Learn More
Systec™ Horizontal, Front-Loading, Benchtop Autoclave, D-100
Systec™ Horizontal, Front-Loading, Benchtop Autoclave, D-100 is compact but designed with an ideal chamber capacity for most standard media bottles and Erlenmeyer flasks.
Brand:  Systec™ 2100
Product Code. 10387552
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
  • Standard features:
    • Housing, support frame and pressure vessel made of corrosion-resistant stainless steel
    • Temperature and pressure range 140°C, 4 bar (except for Systec D-23)
    • LCD display and fully automatic microprocessor control
    • Code-secured access rights for changing parameters and further safety-relevant intervention
    • Flexible PT-100 temperature sensor
  • Optional: Integrated printer for batch documentation
Specifications
Specifications
110 L (Total), 100 L (Nominal)
500 mm
Horizontal, Front-loading Benchtop Autoclave
500 mm
D100
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Systec™ Horizontal, Front-Loading, Benchtop Autoclave, D-100 > D-100 Total/Nominal Capacity: 110/100L