Learn More
Systec™ Horizontal, Front-Loading, Benchtop Autoclave, DE Series
Systec™ Horizontal, Front-Loading, Benchtop Autoclave, DE Series is economical, suitable for basic sterilization applications, and can accept limited additional process optimization options.
POA
Product Code | Brand | Capacity (Metric) | Depth (Metric) Chamber | Diameter (Metric) Chamber | Model | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Depth (Metric) Chamber | Diameter (Metric) Chamber | Model | Price | Quantity & Availability | ||
10405373
|
Systec™
1545 |
50 L (Total), 45 L (Nominal) | 500 mm | 344 mm | DE-45 |
POA
Each |
N/A | ||
12072959
|
Systec™
1590 |
95 L (Total), 90 L (Nominal) | 700 mm | 400 mm | DE-90 |
POA
Each |
N/A | ||
12082959
|
Systec™
1600 |
113 L (Total), 100 L (Nominal) | 500 mm | 500 mm | DE-100 |
POA
Each |
N/A | ||
12002969
|
Systec™
1700 |
211 L (Total), 200 L (Nominal) | 1000 mm | 500 mm | DE200 |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
- Standard features:
- Internal heating elements within the autoclave chamber
- Housing, support frame and pressure vessel made of corrosion-resistant stainless steel
- Temperature and pressure range 140°C, 4 bar (except for Systec DE-23)
- LCD display and fully automatic microprocessor control
- Number of sterilization programs: 12
- Code-secured access rights for changing parameters and further safety-relevant intervention
- Internal memory for storing up to 500 sterilization cycles
- Timer for starting programs
- Autofill: automatic demineralized water feed for steam generation
- Flexible PT-100 temperature sensor
- Temperature holding function for liquids after program finish
- Special program for Durham tubes
- Calculation of F0 value
- Special program for waste sterilization with pulsed heat-up for more efficient air exhaust
- Water-cooled steam exhaust, thermostatically controlled
- Programmable automatic door-opening on completion of program
- RS-232 and RS-485 interfaces for external data transmission (network-compatible)
- Additional options for temperature, pressure, cooling, exhaust filtration, and documentation are available.
Specifications
Horizontal, Front-loading Benchtop Autoclave |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Systec™ Horizontal, Front-Loading, Benchtop Autoclave, DE Series