Learn More
HOXB4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22256-1-AP-150UL
This item is not returnable.
View return policy
Description
HOX homeobox genes play a role both in the early stem cell function as well as in later stages of hematopoietic differentiation. HOXB4, specifically, is a homeotic selector gene implicated in self-renewal of definitive HSCs. Expression of HOXB4 in primitive progenitors combined with culture on hematopoietic stroma induced a switch to the definitive HSC phenotype. Activation of WNT signaling in HSCs induced increased expression of HOXB4. Perturbations of HOX gene expression can cause leukemia.Specifications
HOXB4 | |
Polyclonal | |
Unconjugated | |
HOXB4 | |
homeobox B4, Homeobox protein Hox 2.6, Homeobox protein Hox 2F, Homeobox protein Hox B4, HOX 2.6, HOX2, HOX2F, HOXB4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3214 | |
-20°C | |
Liquid |
Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P17483 | |
HOXB4 | |
HOXB4 Fusion Protein Ag17761 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Product Title
HOXB4 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction