Learn More
HOXB4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22256-1-AP-150UL
 View more versions of this product
Product Code. 16881795
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
HOX homeobox genes play a role both in the early stem cell function as well as in later stages of hematopoietic differentiation. HOXB4, specifically, is a homeotic selector gene implicated in self-renewal of definitive HSCs. Expression of HOXB4 in primitive progenitors combined with culture on hematopoietic stroma induced a switch to the definitive HSC phenotype. Activation of WNT signaling in HSCs induced increased expression of HOXB4. Perturbations of HOX gene expression can cause leukemia.
Specifications
Specifications
HOXB4
Polyclonal
Unconjugated
HOXB4
homeobox B4, Homeobox protein Hox 2.6, Homeobox protein Hox 2F, Homeobox protein Hox B4, HOX 2.6, HOX2, HOX2F, HOXB4
Rabbit
Antigen Affinity Chromatography
RUO
3214
-20°C
Liquid
Western Blot
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P17483
HOXB4
HOXB4 Fusion Protein Ag17761
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HOXB4 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated