missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXD4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17780-1-AP-150UL
This item is not returnable.
View return policy
Description
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.Specifications
HOXD4 | |
Polyclonal | |
Unconjugated | |
HOXD4 | |
HHO.C13, homeobox D4, Homeobox protein HHO.C13, Homeobox protein Hox 4B, Homeobox protein Hox 5.1, Homeobox protein Hox D4, Hox 4.2, HOX 5.1, HOX4, HOX4B, HOXD4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3233 | |
-20°C | |
Liquid |
Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P09016 | |
HOXD4 | |
HOXD4 Fusion Protein Ag11134 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HOXD4 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction