Learn More
HPD Mouse anti-Human, Unconjugated, Clone: 3C9G2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67552-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The HPD antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HPD. This antibody recognizes human, mouse, rat, pig antigen. The HPD antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
HPD | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Pig, Rat, Mouse, Human |
ELISA, Immunohistochemistry, Western Blot | |
3C9G2 | |
Unconjugated | |
150 μL | |
3242 | |
IgG2a |