Learn More
HPD Mouse anti-Human, Unconjugated, Clone: 3C9G2, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | HPD |
---|---|
Clone | 3C9G2 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16840246
|
Proteintech
67552-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16830246
|
Proteintech
67552-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The HPD antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HPD. This antibody recognizes human, mouse, rat, pig antigen. The HPD antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
HPD | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
3242 | |
Primary |
3C9G2 | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Human, Mouse, Rat, Pig | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.