Learn More
HPDL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20777-1-AP-150UL
 View more versions of this product
Product Code. 16840345
  • 4652.90 SEK / 150µL
Description
May have dioxygenase activity.
Specifications
Specifications
HPDL
Polyclonal
Unconjugated
HPDL
4 HPPD L, GLOXD1, HPDL, RP4 534D1.1
Rabbit
Antigen Affinity Chromatography
RUO
84842
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96IR7
HPDL
HPDL Fusion Protein Ag14773
150 μL
Primary
Human
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
HPDL Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated