Learn More
HPDL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20777-1-AP-150UL
 View more versions of this product
Product Code. 16840345
  • 4652.90 SEK / 150µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
May have dioxygenase activity.
Specifications
Specifications
HPDL
Polyclonal
Unconjugated
HPDL
4 HPPD L, GLOXD1, HPDL, RP4 534D1.1
Rabbit
Antigen Affinity Chromatography
RUO
84842
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96IR7
HPDL
HPDL Fusion Protein Ag14773
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HPDL Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated