Läs mer
Tocris Bioscience™ HQL 79
GREENER_CHOICE
Produktkod. 18065892
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18065892 10 mg
18043832 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18065892 Leverantör Tocris Bioscience™ Leverantörsnummer 3530/10
Human hematopoietic prostaglandin D synthase (H-PGDS) inhibitor
Orally active and specific inhibitor of human hematopoietic prostaglandin D synthase (H-PGDS) (IC50 = 6 μM). Antiallergic and anti-inflammatory; displays antiasthmatic activity and potent antihistaminic properties in vivo.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 162641-16-9
Molekylformel C22H27N5O
Specifikationer
Kemiskt namn eller material HQL 79
Kvantitet 10 mg
Formel vikt 377.48
Renhet >99%
Molekylvikt (g/mol) 377.492