missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRAS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18295-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to HRAS and does not recognize NRAS and KRAS.
This gene belongs to the Ras oncogene family, whose members are related to the transforming genes of mammalian sarcoma retroviruses. The products encoded by these genes function in signal transduction pathways. These proteins can bind GTP and GDP, and they have intrinsic GTPase activity. This protein undergoes a continuous cycle of de- and re-palmitoylation, which regulates its rapid exchange between the plasma membrane and the Golgi apparatus. Mutations in this gene cause Costello syndrome, a disease characterized by increased growth at the prenatal stage, growth deficiency at the postnatal stage, predisposition to tumor formation, mental retardation, skin and musculoskeletal abnormalities, distinctive facial appearance and cardiovascular abnormalities. Defects in this gene are implicated in a variety of cancers, including bladder cancer, follicular thyroid cancer, and oral squamous cell carcinoma. Multiple transcript variants, which encode different isoforms, have been identified for this gene.Specifications
HRAS | |
Polyclonal | |
Unconjugated | |
HRAS | |
C BAS/HAS, C H RAS, C HA RAS1, CTLO, GTPase HRas, H Ras 1, H RASIDX, Ha Ras, HAMSV, HRAS, HRAS1, K RAS, p21ras, RASH1, Transforming protein p21 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15461, 293621, 3265 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Paraffin) | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P01112, P20171, Q61411 | |
HRAS | |
Peptide | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HRAS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction