Learn More
HRD1/SYVN1 Mouse anti-Human, Unconjugated, Clone: 2C8A7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67488-1-IG-20UL
 View more versions of this product
Product Code. 16840146
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The HRD1/SYVN1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HRD1/SYVN1. This antibody recognizes human, mouse, rat antigen. The HRD1/SYVN1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
HRD1/SYVN1
Monoclonal
1000 ÎĽg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
2C8A7
Unconjugated
20 ÎĽL
84447
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HRD1/SYVN1 Mouse anti-Human, Unconjugated, Clone: 2C8A7, Proteintech > 20 ÎĽL