Learn More
HSD17B1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25334-1-AP-150UL
 View more versions of this product
Product Code. 16873915
  • 4652.90 SEK / 150µL
Description
The HSD17B1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human HSD17B1. This antibody recognizes human antigen. The HSD17B1 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
Specifications
HSD17B1
Polyclonal
Unconjugated
150 μL
3292
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA
200 μg/mL
Rabbit
Primary
Human
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
HSD17B1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 150 μL