missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSDL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15631-1-AP-150UL
This item is not returnable.
View return policy
Specifications
HSDL2 | |
Polyclonal | |
Unconjugated | |
HSDL2 | |
C9orf99, HSDL2, SDR13C1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
313200, 72479, 84263 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q2TPA8, Q4V8F9, Q6YN16 | |
Hsdl2 | |
HSDL2 Fusion Protein Ag7987 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HSDL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction