Learn More
HSFY1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11970-1-AP-20UL
 View more versions of this product
Product Code. 16879393
  • 1645.54 SEK / 20µL
Description
This gene encodes a member of the heat shock factor (HSF) family of transcriptional activators for heat shock proteins. This gene is a candidate gene for azoospermia, since it localizes to a region of chromosome Y that is sometimes deleted in infertile males. The genome has two identical copies of this gene within a palindromic region; this record represents the more centromeric copy. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Specifications
Specifications
HSFY1
Polyclonal
Unconjugated
HSFY1
HSF2 like, HSF2L, HSFY, HSFY1, HSFY2
Rabbit
Antigen Affinity Chromatography
RUO
86614
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.22 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96LI6
HSFY1
HSFY1 Fusion Protein Ag2592
20 μL
Primary
Human
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
HSFY1 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated