missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hsp47 Antibody (1C4-1A6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22400-0.05mg
This item is not returnable.
View return policy
Description
Hsp47 Monoclonal specifically detects Hsp47 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Hsp47 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence | |
(collagen binding protein 1), Arsenic-transactivated protein 3, AsTP3, CBP1, CBP2RA-A47, Cell proliferation-inducing gene 14 protein, Collagen-binding protein, colligen, Colligin, colligin-1, colligin-2, gp46, HSP47serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1), member 2, member 2, (collagen-binding protein 2), PPROM, proliferation-inducing gene 14, rheumatoid arthritis antigen A-47,47 kDa heat shock protein, Rheumatoid arthritis-related antigen RA-A47, serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), serpin H1, serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, SERPINH2colligin | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Detects 47kDa. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 Îş |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1C4-1A6 | |
Unconjugated | |
P50454.2 | |
SERPINH1 | |
Human HSP47, full length | |
0.05 mg | |
Core ESC Like Genes, Stem Cell Markers | |
871 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hsp47 Antibody (1C4-1A6), Novus Biologicals™ > 0.05 mg, Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction