missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP47 Mouse, anti-Human, Unconjugated, Clone: 2A8F6, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | HSP47 |
---|---|
Clone | 2A8F6 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17211536
|
Proteintech
67863-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17201536
|
Proteintech
67863-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The HSP47 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human HSP47. This antibody recognizes human antigen. The HSP47 antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
HSP47 | |
1000 ÎĽg/mL | |
Monoclonal | |
Liquid | |
Human | |
Primary |
2A8F6 | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
871 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HSP47 Mouse, anti-Human, Unconjugated, Clone: 2A8F6, Proteintech