Learn More
HSPA13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12667-2-AP-150UL
 View more versions of this product
Product Code. 16880504
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
STCH encodes a protein that is a member of the heat shock protein 70 family and is found associated with microsomes. Members of this protein family play a role in the processing of cytosolic and secretory proteins, as well as in the removal of denatured or incorrectly-folded proteins. The encoded protein contains an ATPase domain and has been shown to associate with a ubiquitin-like protein.
Specifications
Specifications
HSPA13
Polyclonal
Unconjugated
HSPA13
Heat shock 70 kDa protein 13, HSPA13, STCH
Rabbit
Antigen Affinity Chromatography
RUO
110920, 29734, 6782
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.16 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O35162, P48723, Q8BM72
Hspa13
HSPA13 Fusion Protein Ag3349
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HSPA13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated