Learn More
HSPA13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12667-2-AP-150UL
 View more versions of this product
Product Code. 16880504
  • 4652.90 SEK / 150µL
Description
STCH encodes a protein that is a member of the heat shock protein 70 family and is found associated with microsomes. Members of this protein family play a role in the processing of cytosolic and secretory proteins, as well as in the removal of denatured or incorrectly-folded proteins. The encoded protein contains an ATPase domain and has been shown to associate with a ubiquitin-like protein.
Specifications
Specifications
HSPA13
Polyclonal
Unconjugated
HSPA13
Heat shock 70 kDa protein 13, HSPA13, STCH
Rabbit
Antigen Affinity Chromatography
RUO
110920, 29734, 6782
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.16 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O35162, P48723, Q8BM72
Hspa13
HSPA13 Fusion Protein Ag3349
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
HSPA13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated