missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human ABCD1 Full-length ORF (NP_000024.2, 1 a.a. - 745 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16181441

Abnova™ Human ABCD1 Full-length ORF (NP_000024.2, 1 a.a. - 745 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16181441
25 μg, 25µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16181441

Brand: Abnova™ H00000215P01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the ALD subfamily, which is involved in peroxisomal import of fatty acids and/or fatty acyl-CoAs in the organelle. All known peroxisomal ABC transporters are half transporters which require a partner half transporter molecule to form a functional homodimeric or heterodimeric transporter. This peroxisomal membrane protein is likely involved in the peroxisomal transport or catabolism of very long chain fatty acids. Defects in this gene have been identified as the underlying cause of adrenoleukodystrophy, an X-chromosome recessively inherited demyelinating disorder of the nervous system. [provided by RefSeq]

Sequence: MPVLSRPRPWRGNTLKRTAVLLALAAYGAHKVYPLVRQCLAPARGLQAPAGEPTQEASGVAAAKAGMNRVFLQRLLWLLRLLFPRVLCRETGLLALHSAALVSRTFLSVYVARLDGRLARCIVRKDPRAFGWQLLQWLLIALPATFVNSAIRYLEGQLALSFRSRLVAHAYRLYFSQQTYYRVSNMDGRLRNPDQSLTEDVVAFAASVAHLYSNLTKPLLDVAVTSYTLLRAARSRGAGTAWPSAIAGLVVFLTANVLRAFSPKFGELVAEEARRKGELRYMHSRVVANSEEIAFYGGHEVELALLQRSYQDLASQINLILLERLWYVMLEQFLMKYVWSASGLLMVAVPIITATGYSESDAEAVKKAALEKKEEELVSERTEAFTIARNLLTAAADAIERIMSSYKEVTELAGYTARVHEMFQVFEDVQRCHFKRPRELEDAQAGSGTIGRSGVRVEGPLKIRGQVVDVEQGIICENIPIVTPSGEVVVASLNIRVEEGMHLLITGPNGCGKSSLFRILGGLWPTYGGVLYKPPPQRMFYIPQRPYMSVGSLRDQVIYPDSVEDMQRKGYSEQDLEAILDVVHLHHILQREGGWEAMCDWKDVLSGGEKQRIGMARMFYHRPKYALLDECTSAVSIDVEGKIFQAAKDAGIALLSITHRPSLWKYHTHLLQFDGEGGWKFEKLDSAARLSLTEEKQRLEQQLAGIPKMQRRLQELCQILGEAVAPAHVPAPSPQGPGGLQGAST

Specifications

Accession Number NP_000024.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 215
Molecular Weight (g/mol) 109.3kDa
Name ABCD1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ABC42/ALD/ALDP/AMN
Common Name ABCD1
Gene Symbol ABCD1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human ABCD1 Full-length ORF (NP_000024.2, 1 a.a. - 745 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.