missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ADAM22 Full-length ORF (AAH36029.1, 1 a.a. - 342 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00053616-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16120413

  • 4515.00 SEK / 10µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This gene is highly expressed in the brain and may function as an integrin ligand in the brain. Alternative splicing results in several transcript variants. [provided by RefSeq]

Sequence: MQAAVAVSVPFLLLCVLGTCPPARCGQAGDASLMELEKRKENRFVERQSIVPLRLIYRSGGEDESRHDALDTRVRGDVGGPQLTHVDQASFQVDAFGTSFILDVVLNHDLLSSEYIERHIEHGGKTVEVKGGEHCYYQGHIRGNPDSFVALSTCHGLHGMFYDGNHTYLIEPEENDTTQEDFHFHSVYKSRLFEFSLDDLPSEFQQVNITPSKFILKPRPKRSKRQLRRYPRNVEEETKYIELMIVNDHLMFKKHRLSVVHTNTYAKSVVNMADLIYKDQLKTRIVLVAMETWATDNKFAISENPLITLREFMKYRRDFIKEKSDAVHLFSYVTSVMMYYFF
Specifications

Specifications

AAH36029.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
65.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ADAM22
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
53616
ADAM22 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MDC2/MGC149832
ADAM22
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers