Learn More
Abnova™ Human ADAMTS17 Partial ORF (NP_620688, 543 a.a. - 650 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00170691-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. ADAMTS family members share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene has a high sequence similarity to the protein encoded by ADAMTS19, another family member. The function of this protein has not been determined. [provided by RefSeq]
Sequence: DGDWSPWGAWSMCSRTCGTGARFRQRKCDNPPPGPGGTHCPGASVEHAVCENLPCPKGLPSFRDQQCQAHDRLSPKKKGLLTAVVVDDKPCELYCSPLGKESPLLVADSpecifications
NP_620688 | |
Liquid | |
170691 | |
ADAMTS17 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ16363/FLJ32769 | |
ADAMTS17 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DGDWSPWGAWSMCSRTCGTGARFRQRKCDNPPPGPGGTHCPGASVEHAVCENLPCPKGLPSFRDQQCQAHDRLSPKKKGLLTAVVVDDKPCELYCSPLGKESPLLVAD | |
RUO | |
ADAMTS17 | |
Wheat Germ (in vitro) | |
GST |