missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ADFP Full-length ORF (NP_001113.2, 1 a.a. - 437 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

4630.00 SEK - 7020.00 SEK

Specifications

Accession Number NP_001113.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 123
Molecular Weight (g/mol) 74.5kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16161271
View Documents
Abnova™
H00000123-P01.25ug
25 ÎĽg
7020.00 SEK
25µg
Please to purchase this item. Need a web account? Register with us today!
16151271
View Documents
Abnova™
H00000123-P01.10ug
10 ÎĽg
4630.00 SEK
10µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Adipocyte differentiation-related protein is associated with the globule surface membrane material. This protein is a major constituent of the globule surface. Increase in mRNA levels is one of the earliest indications of adipocyte differentiation [provided by RefSeq]

Sequence: MASVAVDPQPSVVTRVVNLPLVSSTYDLMSSAYLSTKDQYPYLKSVCEMAENGVKTITSVAMTSALPIIQKLEPQIAVANTYACKGLDRIEERLPILNQPSTQIVANAKGAVTGAKDAVTTTVTGAKDSVASTITGVMDKTKGAVTGSVEKTKSVVSGSINTVLGSRMMQLVSSGVENALTKSELLVEQYLPLTEEELEKEAKKVEGFDLVQKPSYYVRLGSLSTKLHSRAYQQALSRVKEAKQKSQQTISQLHSTVHLIEFARKNVYSANQKIQDAQDKLYLSWVEWKRSIGYDDTDESHCAEHIESRTLAIARNLTQQLQTTCHTLLSNIQGVPQNIQDQAKHMGVMAGDIYSVFRNAASFKEVSDSLLTSSKGQLQKMKESLDDVMDYLVNNTPLNWLVGPFYPQLTESQNAQDQGAEMDKSSQETQRSEHKTH
Specifications

Specifications

NP_001113.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
74.5kDa
Glutathione Sepharose 4 Fast Flow
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ADRP/MGC10598
ADFP
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
123
ADFP (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
RUO
ADFP
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human ADFP Full-length ORF (NP_001113.2, 1 a.a. - 437 a.a.) Recombinant Protein with GST-tag at N-terminal

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.