missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human AKAP10 Partial ORF (NP_009133, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16133926

Abnova™ Human AKAP10 Partial ORF (NP_009133, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16133926
10 μg, 10µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16133926

Brand: Abnova™ H00011216Q01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein interacts with both the type I and type II regulatory subunits of PKA; therefore, it is a dual-specific AKAP. This protein is highly enriched in mitochondria. It contains RGS (regulator of G protein signalling) domains, in addition to a PKA-RII subunit-binding domain. The mitochondrial localization and the presence of RGS domains may have important implications for the function of this protein in PKA and G protein signal transduction. [provided by RefSeq]

Sequence: MRGAGPSPRQSPRTLRPDPGPAMSFFRRKVKGKEQEKTSDVKSIKASISVHSPQKSTKNHALLEAAGPSHVAINAISANMDSFSSSRTATLKKQPSHMEA

Specifications

Accession Number NP_009133
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 11216
Molecular Weight (g/mol) 36.74kDa
Name AKAP10 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MRGAGPSPRQSPRTLRPDPGPAMSFFRRKVKGKEQEKTSDVKSIKASISVHSPQKSTKNHALLEAAGPSHVAINAISANMDSFSSSRTATLKKQPSHMEA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias D-AKAP2/MGC9414/PRKA10
Common Name AKAP10
Gene Symbol AKAP10
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human AKAP10 Partial ORF (NP_009133, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.