missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human AKAP11 Partial ORF (NP_057332, 1801 a.a. - 1901 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16176382

Abnova™ Human AKAP11 Partial ORF (NP_057332, 1801 a.a. - 1901 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16176382
10 μg, 10µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16176382

Brand: Abnova™ H00011215Q01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein is expressed at high levels throughout spermatogenesis and in mature sperm. It binds the RI and RII subunits of PKA in testis. It may serve a function in cell cycle control of both somatic cells and germ cells in addition to its putative role in spermatogenesis and sperm function. [provided by RefSeq]

Sequence: EGLGQDGKTLLITNIDMEPCTVDPQLRIILQWLIASEAEVAELYFHDSANKEFMLLSKQLQEKGWKVGDLLQAVLQYYEVMEKASSEERCKSLFDWLLENA

Specifications

Accession Number NP_057332
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 11215
Molecular Weight (g/mol) 36.85kDa
Name AKAP11 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen EGLGQDGKTLLITNIDMEPCTVDPQLRIILQWLIASEAEVAELYFHDSANKEFMLLSKQLQEKGWKVGDLLQAVLQYYEVMEKASSEERCKSLFDWLLENA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias AKAP220/DKFZp781I12161/FLJ11304/KIAA0629/PRKA11
Common Name AKAP11
Gene Symbol AKAP11
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human AKAP11 Partial ORF (NP_057332, 1801 a.a. - 1901 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.