missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ANTXR2 Partial ORF (NP_477520.2, 241 a.a. - 310 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
This gene encodes a receptor for anthrax toxin. The protein binds to collagen IV and laminin, suggesting that it may be involved in extracellular matrix adhesion. Mutations in this gene cause juvenile hyaline fibromatosis and infantile systemic hyalinosis. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: GRGFMLGSRNGSVLCTYTVNETYTTSVKPVSVQLNSMLCPAPILNKAGETLDVSVSFNGGKSVISGSLIV
Specifications
Specifications
Accession Number | NP_477520.2 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 118429 |
Molecular Weight (g/mol) | 33.44kDa |
Name | ANTXR2 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 25 μg |
Immunogen | GRGFMLGSRNGSVLCTYTVNETYTTSVKPVSVQLNSMLCPAPILNKAGETLDVSVSFNGGKSVISGSLIV |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ Human ANTXR2 Partial ORF (NP_477520.2, 241 a.a. - 310 a.a.) Recombinant Protein with GST-tag at N-terminal >
Spot an opportunity for improvement?Share a Content Correction