Learn More
Abnova™ Human ASB5 Partial ORF (NP_543150, 220 a.a. - 328 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00140458-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. They contain ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Multiple alternatively spliced transcript variants have been described for this gene but their full length sequences are not known. [provided by RefSeq]
Sequence: LLYAGADVQKGKYWDTPLHAAAQQSSTEIVNLLLEFGADINAKNTELLRPIDVATSSSMVERILLQHEATPSSLYQLCRLCIRSYIGKPRLHLIPQLQLPTLLKNFLQYSpecifications
NP_543150 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LLYAGADVQKGKYWDTPLHAAAQQSSTEIVNLLLEFGADINAKNTELLRPIDVATSSSMVERILLQHEATPSSLYQLCRLCIRSYIGKPRLHLIPQLQLPTLLKNFLQY | |
RUO | |
ASB5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
140458 | |
ASB5 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ASB5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |