missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human B3GALT6 Partial ORF (NP_542172, 229 a.a. - 329 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00126792-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16141727

  • 6845.00 SEK / 25µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: RLSRDYLRAWHSEDVSLGAWLAPVDVQREHDPRFDTEYRSRGCSNQYLVTHKQSLEDMLEKHATLAREGRLCKREVQLRLSYVYDWSAPPSQCCQRREGIP
Specifications

Specifications

NP_542172
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.85kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RLSRDYLRAWHSEDVSLGAWLAPVDVQREHDPRFDTEYRSRGCSNQYLVTHKQSLEDMLEKHATLAREGRLCKREVQLRLSYVYDWSAPPSQCCQRREGIP
RUO
B3GALT6
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
126792
B3GALT6 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
beta3GalT6
B3GALT6
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers