missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human B3GALT6 Partial ORF (NP_542172, 229 a.a. - 329 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_542172 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 126792 |
Molecular Weight (g/mol) | 36.85kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16131727
|
Abnova™
H00126792-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16141727
|
Abnova™
H00126792-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sequence: RLSRDYLRAWHSEDVSLGAWLAPVDVQREHDPRFDTEYRSRGCSNQYLVTHKQSLEDMLEKHATLAREGRLCKREVQLRLSYVYDWSAPPSQCCQRREGIPSpecifications
NP_542172 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.85kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
beta3GalT6 | |
B3GALT6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
126792 | |
B3GALT6 (Human) Recombinant Protein (Q01) | |
RLSRDYLRAWHSEDVSLGAWLAPVDVQREHDPRFDTEYRSRGCSNQYLVTHKQSLEDMLEKHATLAREGRLCKREVQLRLSYVYDWSAPPSQCCQRREGIP | |
RUO | |
B3GALT6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |