missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human B3GNT7 Full-length ORF (AAI48681.1, 1 a.a. - 401 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
Sequence: MSLWKKTVYRSLCLALALLVAVTVFQRSLTPGQFLQEPPPPTLEPQKAQKPNGQLVNPNNFWKNPKDVAAPTPMASQGPQAWDVTTTNCSANINLTHQPWFQVLEPQFRQFLFYRHCRYFPMLLNHPEKCRGDVYLLVVVKSVITQHDRREAIRQTWGRERQSAGGGRGAVRTLFLLGTASKQEERTHYQQLLAYEDRLYGDILQWGFLDTFFNLTLKEIHFLKWLDIYCPHVPFIFKGDDDVFVNPTNLLEFLADRQPQENLFVGDVLQHARPIRRKDNKYYIPGALYGKASYPPYAGGGGFLMAGSLARRLHHACDTLELYPIDDVFLGMCLEVLGVQPTAHEGFKTFGISRNRNSRMNKEPCFFRAMLVVHKLLPPELLAMWGLVHSNLTCSRKLQVL
Specifications
Specifications
Accession Number | AAI48681.1 |
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 93010 |
Molecular Weight (g/mol) | 71.06kDa |
Name | B3GNT7 (Human) Recombinant Protein (P01) |
Purification Method | Glutathione Sepharose 4 Fast Flow |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 10 μg |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Safety and Handling
Product Identifier
- B3GNT7 (Human) Recombinant Protein (P01)
- MIXTURE LIST-Contains : tris-HCl, reduced glutathione
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ Human B3GNT7 Full-length ORF (AAI48681.1, 1 a.a. - 401 a.a.) Recombinant Protein with GST-tag at N-terminal >
Spot an opportunity for improvement?Share a Content Correction