Learn More
Abnova™ Human BCAS1 Full-length ORF (ADR83077.1, 1 a.a. - 584 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008537-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene resides in a region at 20q13 which is amplified in a variety of tumor types and associated with more aggressive tumor phenotypes. Among the genes identified from this region, it was found to be highly expressed in three amplified breast cancer cell lines and in one breast tumor without amplification at 20q13.2. However, this gene is not in the common region of maximal amplification and its expression was not detected in the breast cancer cell line MCF7, in which this region is highly amplified. Although not consistently expressed, this gene is a candidate oncogene. [provided by RefSeq]
Sequence: MGNQMSVPQRVEDQENEPEAETYKDNASALNGVPVVVSTHTVQHLEEVDLGISVKTDNVATSSPETTEISAVADANGKNLGKEAKPEAPAAKSRFFLMLSRPVPGRTGDQAADSSLGSVKLDVSSNKAPANKDPSESWTLPVAAGPGQDTDKTPGHAPAQDKALSAARDPTLLPPETGGAGGEAPSKPKDSSFFDKFFKLDKGQEKVPGDSQQEAKRAEHQDKVDEVPGLSGQSDDVPAGKDIVDGKEKEGQELGTADCSVPGDPEGLETAKDDSQAAAIAENNNSIMSFFKTLVSPNKAETKKDPEDTGAEKSPTTSADLKSDKANFTSQETQGAGKNSKGCNPSGHTQSVTTPEPAKEGTKEKSGPTSLPLGKLFWKKSVKEDSVPTGAEENVVCESPVEIIKSKEVESALQTVDLNEGDAAPEPTEAKLKREESKPRTSLMAFLRQMSVKGDGGITHSEEINGKDSSCQTSDSTEKTITPPEPEPTGAPQKGKEGSSKDKKSAAEMNKQKSNKQEAKEPAQCTEQATVDTNSLQNGDKLQKRPEKRQQSLGGFFKGLGPKRMLDAQVQTDPVSIGPVGKSKSpecifications
ADR83077.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
90.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
BCAS1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
8537 | |
BCAS1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AIBC1/NABC1 | |
BCAS1 | |
Recombinant | |
wheat germ expression system |