Learn More
Abnova™ Human BIRC6 Partial ORF (NP_057336, 31 a.a. - 140 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00057448-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein with a BIR (baculoviral inhibition of apoptosis protein repeat) domain and a UBCc (ubiquitin-conjugating enzyme E2, catalytic) domain. This protein inhibits apoptosis by facilitating the degradation of apoptotic proteins by ubiquitination. [provided by RefSeq]
Sequence: RDGCMHCDADGLHSLSYHPALNAILAVTSRGTIKVIDGTSGATLQASALSAKPGGQVKCQYISAVDKVIFVDDYAVGCRKDLNGILLLDTALQTPVSKQDDVVQLELPVTSpecifications
NP_057336 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RDGCMHCDADGLHSLSYHPALNAILAVTSRGTIKVIDGTSGATLQASALSAKPGGQVKCQYISAVDKVIFVDDYAVGCRKDLNGILLLDTALQTPVSKQDDVVQLELPVT | |
RUO | |
BIRC6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
57448 | |
BIRC6 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APOLLON/BRUCE/FLJ13726/FLJ13786/KIAA1289 | |
BIRC6 | |
Recombinant | |
wheat germ expression system |