missing translation for 'onlineSavingsMsg'
Learn More
Invitrogen™ Human BRCA2 (aa 1255-1385) Control Fragment Recombinant Protein Product Code.: 30195359

Invitrogen™ Human BRCA2 (aa 1255-1385) Control Fragment Recombinant Protein

Product Code. 30195359
100 μL, 100µL
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195359

Brand: Invitrogen™ RP89275

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (50%), Rat (50%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82936 (PA5-82936. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Breast Cancer Type 2 susceptibility protein (BRCA2) is a tumor-suppressor protein reported to be involved in double-strand break repair, homologous recombination and S-phase checkpoint activation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P51587
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 675
Name Human BRCA2 (aa 1255-1385) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias brca 2; BRCA1/BRCA2-containing complex, subunit 2; BRCA2; brca-2; BRCA2 DNA repair associated; BRCA2, DNA repair associated; BRCC2; breast and ovarian cancer susceptibility gene, early onset; breast and ovarian cancer susceptibility protein 2; breast cancer 2; breast cancer 2 tumor suppressor; breast cancer 2, early onset; breast cancer 2, mutation 1, University of Wisconsin-Madison; breast cancer susceptibility protein 2; breast cancer type 2 susceptibility protein; Breast cancer type 2 susceptibility protein homolog; BROVCA2; FACD; FAD; FAD1; FANCB; FANCD; FANCD1; Fanconi anemia group D1 protein; Fanconi anemia group D1 protein homolog; GLM3; mutant BRCA2; PNCA2; RAB163; RP11-298P3.4; XRCC11
Common Name BRCA2
Gene Symbol BRCA2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VHPISLSSSKCHDSVVSMFKIENHNDKTVSEKNNKCQLILQNNIEMTTGTFVEEITENYKRNTENEDNKYTAASRNSHNLEFDGSDSSKNDTVCIHKDETDLLFTDQHNICLKLSGQFMKEGNTQIKEDLS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Human BRCA2 (aa 1255-1385) Control Fragment Recombinant Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.