missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BTBD1 Partial ORF (NP_079514, 384 a.a. - 482 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00053339-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16147366

  • 6845.00 SEK / 25µg
Estimated Shipment: 10-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The C-terminus of the protein encoded by this gene binds topoisomerase I. The N-terminus contains a proline-rich region and a BTB/POZ domain (broad-complex, Tramtrack and bric a brac/Pox virus and Zinc finger), both of which are typically involved in protein-protein interactions. Subcellularly, the protein localizes to cytoplasmic bodies. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: EYEKKQTLGQNDTGFSCDGTANTFRVMFKEPIEILPNVCYTACATLKGPDSHYGTKGLKKVVHETPAASKTVFFFFSSPGNNNGTSIEDGQIPEIIFYT
Specifications

Specifications

NP_079514
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EYEKKQTLGQNDTGFSCDGTANTFRVMFKEPIEILPNVCYTACATLKGPDSHYGTKGLKKVVHETPAASKTVFFFFSSPGNNNGTSIEDGQIPEIIFYT
RUO
BTBD1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
53339
BTBD1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C15orf1/NS5ATP8
BTBD1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers