missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human BTBD1 Partial ORF (NP_079514, 384 a.a. - 482 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_079514 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 53339 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16137366
|
Abnova™
H00053339-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16147366
|
Abnova™
H00053339-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The C-terminus of the protein encoded by this gene binds topoisomerase I. The N-terminus contains a proline-rich region and a BTB/POZ domain (broad-complex, Tramtrack and bric a brac/Pox virus and Zinc finger), both of which are typically involved in protein-protein interactions. Subcellularly, the protein localizes to cytoplasmic bodies. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: EYEKKQTLGQNDTGFSCDGTANTFRVMFKEPIEILPNVCYTACATLKGPDSHYGTKGLKKVVHETPAASKTVFFFFSSPGNNNGTSIEDGQIPEIIFYTSpecifications
NP_079514 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C15orf1/NS5ATP8 | |
BTBD1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
53339 | |
BTBD1 (Human) Recombinant Protein (Q01) | |
EYEKKQTLGQNDTGFSCDGTANTFRVMFKEPIEILPNVCYTACATLKGPDSHYGTKGLKKVVHETPAASKTVFFFFSSPGNNNGTSIEDGQIPEIIFYT | |
RUO | |
BTBD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |