missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human C11orf16 Full-length ORF (AAH27865.1, 1 a.a. - 404 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
Sequence: MESSTGPRMPLLKYCSVATSLKAPGWDGAAPPWDLSFTYPFALQAPWLTGHKPLARHASSCPCLHVADPAWQGPGWLGRAGDAANTWVLARREADGFYYRAQIKATPELERQGVLLVEFEAPLVAGPKLPAQQQRVVLEEDVIPLSPSVGYSLRPGDKVLALWEPGQQQYGPGTVLLGLEMRDPQRASKEKEITVHFWNGKAAKVPLGGVQSVSLTIWKKAVERLHKSFTREHPRPLHWAPCCSLLGPITGRITNELPPDAPFLCPLCHHHACCQLLCQGCLCGCPPCGTTWWPLTRTSEVMARELPELEATAQLLPLEGPKEEKVAMHAPLAVSSSSSSSCEQDGVENDLEMGPPQRLMVNSAVNTDPIFLEMPLRQSGLCQPEWRYWKRNGPEPCPGKPGKE
Specifications
Specifications
Accession Number | AAH27865.1 |
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 56673 |
Molecular Weight (g/mol) | 70.8kDa |
Name | C11orf16 (Human) Recombinant Protein (P01) |
Purification Method | Glutathione Sepharose 4 Fast Flow |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue |
Quantity | 25 μg |
Immunogen | MESSTGPRMPLLKYCSVATSLKAPGWDGAAPPWDLSFTYPFALQAPWLTGHKPLARHASSCPCLHVADPAWQGPGWLGRAGDAANTWVLARREADGFYYRAQIKATPELERQGVLLVEFEAPLVAGPKLPAQQQRVVLEEDVIPLSPSVGYSLRPGDKVLALWEPGQQQYGPGTVLLGLEMRDPQRASKEKEITVHFWNGKAAKVPLGGVQSVSLTIWKKAVERLHKSFTREHPRPLHWAPCCSLLGPITGRITNELPPDAPFLCPLCHHHACCQLLCQGCLCGCPPCGTTWWPLTRTSEVMARELPELEATAQLLPLEGPKEEKVAMHAPLAVSSSSSSSCEQDGVENDLEMGPPQRLMVNSAVNTDPIFLEMPLRQSGLCQPEWRYWKRNGPEPCPGKPGKE |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ Human C11orf16 Full-length ORF (AAH27865.1, 1 a.a. - 404 a.a.) Recombinant Protein with GST-tag at N-terminal >
Spot an opportunity for improvement?Share a Content Correction