missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human C9orf12 Partial ORF (NP_073592, 391 a.a. - 491 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00064768-Q01.10ug
This item is not returnable.
View return policy
Description
Sequence: VQQYRVAMTAKDCSIMIALSPCLQDASSDQRPVVPSSRSRFAFSVSVLDLDLKPYESIPHQYKLDGKIVNYYSKTVRAKDNAVMSTRFKESEDCTLVLHKVSpecifications
NP_073592 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.85kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VQQYRVAMTAKDCSIMIALSPCLQDASSDQRPVVPSSRSRFAFSVSVLDLDLKPYESIPHQYKLDGKIVNYYSKTVRAKDNAVMSTRFKESEDCTLVLHKV | |
RUO | |
IPPK | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
64768 | |
C9orf12 (Human) Recombinant Protein (Q01) | |
10 ÎĽg | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C9orf12/FLJ13163/INSP5K2/IP5K/IPK1/KIAA0699/bA476B13.1 | |
IPPK | |
Recombinant | |
wheat germ expression system |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ Human C9orf12 Partial ORF (NP_073592, 391 a.a. - 491 a.a.) Recombinant Protein with GST-tag at N-terminal > 10μg
Spot an opportunity for improvement?Share a Content Correction