Learn More
Abnova™ Human Ca1 Partial Orf (Aah27890.1, 12 A.A.-150 A.A.) Recombinant Protein With Gst Tag At N-Terminal.
Recombinant proteins expressed from in vitro wheat germ system. Such proteins mirror the conformation and folding of the native proteins in the eukaryotic biological system.
Brand: Abnova™ H00000759-Q01.10ug
Additional Details : Weight : 0.02000kg
Description
- Good Folding
- Excellent amino acids labeling
Immunogen Sequence: NGPEQWSKLYPIANGNNQSPVDIKTSETKHDTSLKPISVSYNPATAKEIINVGHSFHVNFEDNDNRSVLKGGPFSDSYRLFQFHFHWGSTNEHGSEHTVDGVKYSAELHVAHWNSAKYSSLAEAASKADGLAVIGVLMK
Specifications
AAH27890.1 | |
Liquid | |
759 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Car1 | |
CA1 |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer. | |
CA1 (Human) Recombinant Protein (Q01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
NGPEQWSKLYPIANGNNQSPVDIKTSETKHDTSLKPISVSYNPATAKEIINVGHSFHVNFEDNDNRSVLKGGPFSDSYRLFQFHFHWGSTNEHGSEHTVDGVKYSAELHVAHWNSAKYSSLAEAASKADGLAVIGVLMK | |
RUO | |
CA1 | |
GST |
For Research Use Only