missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human CACNA1S Partial ORF (NP_000060, 1743 a.a. - 1838 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16185504

Abnova™ Human CACNA1S Partial ORF (NP_000060, 1743 a.a. - 1838 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16185504
10 ug, 10µg
Click to view available options
Quantity:
10 ug
25 ug
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16185504

Brand: Abnova™ H00000779Q01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

This gene encodes one of the five subunits of the slowly inactivating L-type voltage-dependent calcium channel in skeletal muscle cells. Mutations in this gene have been associated with hypokalemic periodic paralysis, thyrotoxic periodic paralysis and malignant hyperthermia susceptibility. [provided by RefSeq]

Sequence: CPRVESSMPEDRKSSTPGSLHEETPHSRSTRENTSRCSAPATALLIQKALVRGGLGTLAADANFIMATGQALGDACQMEPEEVEIMATELLKGREA

Specifications

Accession Number NP_000060
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 779
Molecular Weight (g/mol) 36.3kDa
Name CACNA1S (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen CPRVESSMPEDRKSSTPGSLHEETPHSRSTRENTSRCSAPATALLIQKALVRGGLGTLAADANFIMATGQALGDACQMEPEEVEIMATELLKGREA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CACNL1A3/CCHL1A3/Cav1.1/HOKPP/MHS5/hypoPP
Common Name CACNA1S
Gene Symbol CACNA1S
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human CACNA1S Partial ORF (NP_000060, 1743 a.a. - 1838 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.