missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CASC4 Full-length ORF (NP_612432.2, 1 a.a. - 436 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16176983
25 μg, 25µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy
This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

The increased expression level of this gene is associated with HER-2/neu proto-oncogene overexpression. Amplification and resulting overexpression of this proto-oncogene are found in approximately 30% of human breast and 20% of human ovarian cancers. Further characterization of the product of this gene may yield insight into the fundamental biology and pathogenetic effects of HER-2/neu overexpression in human breast and ovarian cancer cells. Alternatively spliced variants encoding different isoforms have been identified for this gene. [provided by RefSeq]

Sequence: MVGFGANRRAGRLPSLVLVVLLVVIVVLAFNYWSISSRHVLLQEEVAELQGQVQRTEVARGRLEKRNSDLLLLVDTHKKQIDQKEADYGRLSSRLQAREGLGKRCEDDKVKLQNNISYQMADIHHLKEQLAELRQEFLRQEDQLQDYRKNNTYLVKRLEYESFQCGQQMKELRAQHEENIKKLADQFLEEQKQETQKIQSNDGKELDINNQVVPKNIPKVAENVADKNEEPSSNHIPHGKEQIKRGGDAGMPGIEENDLAKVDDLPPALRKPPISVSQHESHQAISHLPTGQPLSPNMPPDSHINHNGNPGTSKQNPSSPLQRLIPGSNLDSEPRIQTDILKQATKDRVSDFHKLKQSRFFDENESPVDPQHGSKLADYNGDDGNVGEYEADKQAELAYNEEEDGDGGEEDVQDDEERELQMDPADYGKQHFNDVL

Specifications

Accession Number NP_612432.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 113201
Molecular Weight (g/mol) 75.9kDa
Name CASC4 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MVGFGANRRAGRLPSLVLVVLLVVIVVLAFNYWSISSRHVLLQEEVAELQGQVQRTEVARGRLEKRNSDLLLLVDTHKKQIDQKEADYGRLSSRLQAREGLGKRCEDDKVKLQNNISYQMADIHHLKEQLAELRQEFLRQEDQLQDYRKNNTYLVKRLEYESFQCGQQMKELRAQHEENIKKLADQFLEEQKQETQKIQSNDGKELDINNQVVPKNIPKVAENVADKNEEPSSNHIPHGKEQIKRGGDAGMPGIEENDLAKVDDLPPALRKPPISVSQHESHQAISHLPTGQPLSPNMPPDSHINHNGNPGTSKQNPSSPLQRLIPGSNLDSEPRIQTDILKQATKDRVSDFHKLKQSRFFDENESPVDPQHGSKLADYNGDDGNVGEYEADKQAELAYNEEEDGDGGEEDVQDDEERELQMDPADYGKQHFNDVL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp459F1927/H63/MGC74708
Common Name CASC4
Gene Symbol CASC4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human CASC4 Full-length ORF (NP_612432.2, 1 a.a. - 436 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.