Learn More
Abnova™ Human CDC42EP2 Partial ORF (NP_006770, 102 a.a. - 210 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
CDC42, a small Rho GTPase, regulates the formation of F-actin-containing structures through its interaction with the downstream effector proteins. The protein encoded by this gene is a member of the Borg family of CDC42 effector proteins. Borg family proteins contain a CRIB (Cdc42/Rac interactive-binding) domain. They bind to, and negatively regulate the function of, CDC42. Coexpression of this protein with dominant negative mutant CDC42 protein in fibroblast was found to induce pseudopodia formation, which suggested a role of this protein in actin filament assembly and cell shape control. [provided by RefSeq]
Specifications
Specifications
Accession Number | NP_006770 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10435 |
Molecular Weight (g/mol) | 37.73kDa |
Name | CDC42EP2 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 25 μg |
Immunogen | PLLKNAISLPVIGGPQALTLPTAQAPPKPPRLHLETPQPSPQEGGSVDIWRIPETGSPNSGLTPESGAEEPFLSNASSLLSLHVDLGPSILDDVLQIMDQDLDSMQIPT |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.