missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CDON Partial ORF (NP_058648, 1155 a.a. - 1263 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
CDON and BOC (MIM 608708) are cell surface receptors of the immunoglobulin (Ig)/fibronectin type III (FNIII; see MIM 135600) repeat family involved in myogenic differentiation. CDON and BOC are coexpressed during development, form complexes with each other in a cis fashion, and are related to each other in their ectodomains, but each has a unique long cytoplasmic tail.[supplied by OMIM]
Sequence: VKVPVCLTSAVPDCGQLPEESVKDNVEPVPTQRTCCQDIVNDVSSDGSEDPAEFSRGDSCAHSETEINIVSWNALILPPVPEGCAEKTMWSPPGIPLDSPTEVLQQPRE
Specifications
Specifications
Accession Number | NP_058648 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 50937 |
Molecular Weight (g/mol) | 37.73kDa |
Name | CDON (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 25 μg |
Immunogen | VKVPVCLTSAVPDCGQLPEESVKDNVEPVPTQRTCCQDIVNDVSSDGSEDPAEFSRGDSCAHSETEINIVSWNALILPPVPEGCAEKTMWSPPGIPLDSPTEVLQQPRE |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ Human CDON Partial ORF (NP_058648, 1155 a.a. - 1263 a.a.) Recombinant Protein with GST-tag at N-terminal >
Spot an opportunity for improvement?Share a Content Correction