missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CHST12 Partial ORF (NP_061111, 183 a.a. - 292 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
Chondroitin 4-sulfotransferases, such as CHST12, add sulfate to position 4 of N-acetylgalactosamine residues in chondroitin (Hiraoka et al., 2000 [PubMed 10781601]).[supplied by OMIM]
Sequence: IVLSGSLLHRGAPYRDPLRIPREHVHNASAHLTFNKFWRRYGKLSRHLMKVKLKKYTKFLFVRDPFVRLISAFRSKFELENEEFYRKFAVPMLRLYANHTSLPASAREAF
Specifications
Specifications
Accession Number | NP_061111 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 55501 |
Molecular Weight (g/mol) | 37.84kDa |
Name | CHST12 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 25 μg |
Immunogen | IVLSGSLLHRGAPYRDPLRIPREHVHNASAHLTFNKFWRRYGKLSRHLMKVKLKKYTKFLFVRDPFVRLISAFRSKFELENEEFYRKFAVPMLRLYANHTSLPASAREAF |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ Human CHST12 Partial ORF (NP_061111, 183 a.a. - 292 a.a.) Recombinant Protein with GST-tag at N-terminal >
Spot an opportunity for improvement?Share a Content Correction