missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CLDN7 Full-length ORF (NP_001298.2, 1 a.a. - 211 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

4125.00 SEK - 6260.00 SEK

Specifications

Accession Number NP_001298.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 1366
Molecular Weight (g/mol) 48.8kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16146637
View Documents Promotion Details
Abnova™
H00001366-P01.25UG
25 ug
6260.00 SEK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 06-11-2023
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16136637
View Documents Promotion Details
Abnova™
H00001366-P01.10UG
10 ug
4125.00 SEK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 06-11-2023
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Claudins, such as CLDN7, are involved in the formation of tight junctions between epithelial cells. Tight junctions restrict lateral diffusion of lipids and membrane proteins, and thereby physically define the border between the apical and basolateral compartments of epithelial cells (Zheng et al., 2003 [PubMed 14502431]).[supplied by OMIM]

Sequence: MANSGLQLLGFSMALLGWVGLVACTAIPQWQMSSYAGDNIITAQAMYKGLWMDCVTQSTGMMSCKMYDSVLALSAALQATRALMVVSLVLGFLAMFVATMGMKCTRCGGDDKVKKARIAMGGGIIFIVAGLATLVACSWYGHQIVTDFYNPLIPTNIKYEFGPAIFIGWAGSALVILGGALLSCSCPGNESKAGYRAPRSYPKSNSSKEYV
Specifications

Specifications

NP_001298.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
48.8kDa
Glutathione Sepharose 4 Fast Flow
MANSGLQLLGFSMALLGWVGLVACTAIPQWQMSSYAGDNIITAQAMYKGLWMDCVTQSTGMMSCKMYDSVLALSAALQATRALMVVSLVLGFLAMFVATMGMKCTRCGGDDKVKKARIAMGGGIIFIVAGLATLVACSWYGHQIVTDFYNPLIPTNIKYEFGPAIFIGWAGSALVILGGALLSCSCPGNESKAGYRAPRSYPKSNSSKEYV
RUO
CLDN7
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
1366
CLDN7 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CEPTRL2/CPETRL2/Hs.84359/claudin-1
CLDN7
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Product Certifications