Learn More
Abnova™ Human CNO Partial ORF (NP_060836, 108 a.a. - 217 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
This intronless gene encodes a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. A similar protein in mouse is a component of a protein complex termed biogenesis of lysosome-related organelles complex 1 (BLOC-1), and is a model for Hermansky-Pudlak syndrome. The encoded protein may play a role in intracellular vesicular trafficking. [provided by RefSeq]
Specifications
Specifications
Accession Number | NP_060836 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 55330 |
Molecular Weight (g/mol) | 37.84kDa |
Name | CNO (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 25 μg |
Immunogen | GDSSHVVSEGVPRIHAKAAEMRRIYSRIDRLEAFVRMVGGRVARMEEQVTKAEAELGTFPRAFKKLLHTMNVPSLFSKSAPSRPQQAGYEAPVLFRTEDYFPCCSERPQL |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.