missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CTF1 Full-length ORF (NP_001321.1, 1 a.a. - 201 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantity:
10 ug
25 ug
Unit Size:
10µg
25µg
Description
The protein encoded by this gene is a secreted cytokine that induces cardiac myocyte hypertrophy in vitro. It has been shown to bind and activate the ILST/gp130 receoptor. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MSRREGSLEDPQTDSSVSLLPHLEAKIRQTHSLAHLLTKYAEQLLQEYVQLQGDPFGLPSFSPPRLPVAGLSAPAPSHAGLPVHERLRLDAAALAALPPLLDAVCRRQAELNPRAPRLLRRLEDAARQARALGAAVEALLAALGAANRGPRAEPPAATASAASATGVFPAKVLGLRVCGLYREWLSRTEGDLGQLLPGGSA
Specifications
Specifications
Accession Number | NP_001321.1 |
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1489 |
Molecular Weight (g/mol) | 47.6kDa |
Name | CTF1 (Human) Recombinant Protein (P01) |
Purification Method | Glutathione Sepharose 4 Fast Flow |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue |
Quantity | 25 ug |
Immunogen | MSRREGSLEDPQTDSSVSLLPHLEAKIRQTHSLAHLLTKYAEQLLQEYVQLQGDPFGLPSFSPPRLPVAGLSAPAPSHAGLPVHERLRLDAAALAALPPLLDAVCRRQAELNPRAPRLLRRLEDAARQARALGAAVEALLAALGAANRGPRAEPPAATASAASATGVFPAKVLGLRVCGLYREWLSRTEGDLGQLLPGGSA |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ Human CTF1 Full-length ORF (NP_001321.1, 1 a.a. - 201 a.a.) Recombinant Protein with GST-tag at N-terminal >
Spot an opportunity for improvement?Share a Content Correction