Learn More
Abnova™ Human DDEFL1 Full-length ORF (NP_060177.2, 1 a.a. - 903 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055616-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of a subfamily of ADP-ribosylation factor(Arf) GTPase-activating proteins that contain additional ankyrin repeat and pleckstrin homology domains. The Arf GAP domain of this protein catalyzes the hydrolysis of GTP bound to Arf proteins. The encoded protein promotes cell differentiation and migration and has been implicated in cancer cell invasion. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: MPEQFSVAEFLAVTAEDLSSPAGAAAFAAKMPRYRGAALAREEILEGDQAILQRIKKAVRAIHSSGLGHVENEEQYREAVESLGNSHLSQNSHELSTGFLNLAVFTREVAALFKNLIQNLNNIVSFPLDSLMKGQLRDGRQDSKKQLEKAWKDYEAKMAKLEKERDRARVTGGIPGEVAQDMQRERRIFQLHMCEYLLKAGESQMKQGPDFLQSLIKFFHAQHNFFQDGWKAAQSLFPFIEKLAASVHALHQAQEDELQKLTQLRDSLRGTLQLESREEHLSRKNSGCGYSIHQHQGNKQFGTEKVGFLYKKSDGIRRVWQKRKCGVKYGCLTISHSTINRPPVKLTLLTCQVRPNPEEKKCFDLVTHNRTYHFQAEDEHECEAWVSVLQNSKDEALSSAFLGEPSAGPGSWGSAGHDGEPHDLTKLLIAEVKSRPGNSQCCDCGAADPTWLSTNLGVLTCIQCSGVHRELGVRFSRMQSLTLDLLGPSELLLALNMGNTSFNEVMEAQLPSHGGPKPSAESDMGTRRDYIMAKYVEHRFARRCTPEPQRLWTAICNRDLLSVLEAFANGQDFGQPLPGPDAQAPEELVLHLAVKVANQASLPLVDFIIQNGGHLDAKAADGNTALHYAALYNQPDCLKLLLKGRALVGTVNEAGETALDIARKKHHKECEELLEQAQAGTFAFPLHVDYSWVISTEPGSDSEEDEEEKRCLLKLPAQAHWASGRLDISNKTYETVASLGAATPQGESEDCPPPLPVKNSSRTLVQGCARHASGDRSEVSSLSSEAPETPESLGSPASSSSLMSPLEPGDPSQAPPNSEEGLREPPGTSRPSLTSGTTPSEMYLPVRFSSESTRSYRRGARSPEDGPSARQPLPRRNVPVGITEGDGSRTGSLPASSVQLLQDSpecifications
NP_060177.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
125.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ASAP3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
55616 | |
DDEFL1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACAP4/CENTB6/DDEFL1/FLJ20199/UPLC1 | |
ASAP3 | |
Recombinant | |
wheat germ expression system |