missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DDEFL1 Full-length ORF (NP_060177.2, 1 a.a. - 903 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_060177.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 55616 |
Molecular Weight (g/mol) | 125.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16141603
|
Abnova™
H00055616-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16131603
|
Abnova™
H00055616-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of a subfamily of ADP-ribosylation factor(Arf) GTPase-activating proteins that contain additional ankyrin repeat and pleckstrin homology domains. The Arf GAP domain of this protein catalyzes the hydrolysis of GTP bound to Arf proteins. The encoded protein promotes cell differentiation and migration and has been implicated in cancer cell invasion. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: MPEQFSVAEFLAVTAEDLSSPAGAAAFAAKMPRYRGAALAREEILEGDQAILQRIKKAVRAIHSSGLGHVENEEQYREAVESLGNSHLSQNSHELSTGFLNLAVFTREVAALFKNLIQNLNNIVSFPLDSLMKGQLRDGRQDSKKQLEKAWKDYEAKMAKLEKERDRARVTGGIPGEVAQDMQRERRIFQLHMCEYLLKAGESQMKQGPDFLQSLIKFFHAQHNFFQDGWKAAQSLFPFIEKLAASVHALHQAQEDELQKLTQLRDSLRGTLQLESREEHLSRKNSGCGYSIHQHQGNKQFGTEKVGFLYKKSDGIRRVWQKRKCGVKYGCLTISHSTINRPPVKLTLLTCQVRPNPEEKKCFDLVTHNRTYHFQAEDEHECEAWVSVLQNSKDEALSSAFLGEPSAGPGSWGSAGHDGEPHDLTKLLIAEVKSRPGNSQCCDCGAADPTWLSTNLGVLTCIQCSGVHRELGVRFSRMQSLTLDLLGPSELLLALNMGNTSFNEVMEAQLPSHGGPKPSAESDMGTRRDYIMAKYVEHRFARRCTPEPQRLWTAICNRDLLSVLEAFANGQDFGQPLPGPDAQAPEELVLHLAVKVANQASLPLVDFIIQNGGHLDAKAADGNTALHYAALYNQPDCLKLLLKGRALVGTVNEAGETALDIARKKHHKECEELLEQAQAGTFAFPLHVDYSWVISTEPGSDSEEDEEEKRCLLKLPAQAHWASGRLDISNKTYETVASLGAATPQGESEDCPPPLPVKNSSRTLVQGCARHASGDRSEVSSLSSEAPETPESLGSPASSSSLMSPLEPGDPSQAPPNSEEGLREPPGTSRPSLTSGTTPSEMYLPVRFSSESTRSYRRGARSPEDGPSARQPLPRRNVPVGITEGDGSRTGSLPASSVQLLQDSpecifications
NP_060177.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
125.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACAP4/CENTB6/DDEFL1/FLJ20199/UPLC1 | |
ASAP3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
55616 | |
DDEFL1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
ASAP3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |