missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DDO Partial ORF (NP_003640, 270 a.a. - 369 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
The protein encoded by this gene is a peroxisomal flavoprotein that catalyzes the oxidative deamination of D-aspartate and N-methyl D-aspartate. Flavin adenine dinucleotide or 6-hydroxyflavin adenine dinucleotide can serve as the cofactor in this reaction. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: WNLSPDAENSREILSRCCALEPSLHGACNIREKVGLRPYRPGVRLQTELLARDGQRLPVVHHYGHGSGGISVHWGTALEAARLVSECVHALRTPIPKSNL
Specifications
Specifications
Accession Number | NP_003640 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8528 |
Molecular Weight (g/mol) | 36.74kDa |
Name | DDO (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 25 μg |
Immunogen | WNLSPDAENSREILSRCCALEPSLHGACNIREKVGLRPYRPGVRLQTELLARDGQRLPVVHHYGHGSGGISVHWGTALEAARLVSECVHALRTPIPKSNL |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ Human DDO Partial ORF (NP_003640, 270 a.a. - 369 a.a.) Recombinant Protein with GST-tag at N-terminal >
Spot an opportunity for improvement?Share a Content Correction