missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DGKH Partial ORF (AAH43292, 831 a.a. - 930 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
This gene encodes a member of the diacylglycerol kinase (DGK) enzyme family of proteins, specifically the type II DGK subfamily. Members of this family are involved in regulating the intracellular concentrations of diacylglycerol and phosphatidic acid. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: SGKPVLRTHLYIHHAIDLATEEVSQMQLCSQAAEELITRICDAATIHCLLEQELAHAVNACSHALNKANPRCPESLTRDTATEIAINVKALYNETESLLV
Specifications
Specifications
Accession Number | AAH43292 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 160851 |
Molecular Weight (g/mol) | 36.63kDa |
Name | DGKH (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 25 μg |
Immunogen | SGKPVLRTHLYIHHAIDLATEEVSQMQLCSQAAEELITRICDAATIHCLLEQELAHAVNACSHALNKANPRCPESLTRDTATEIAINVKALYNETESLLV |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ Human DGKH Partial ORF (AAH43292, 831 a.a. - 930 a.a.) Recombinant Protein with GST-tag at N-terminal >
Spot an opportunity for improvement?Share a Content Correction